Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ UVA Sensor
SDPSCI_ED

Catalog No. S16272ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
Middle School, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16272ND Middle School, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16272ND Supplier Vernier™ Supplier No. UVABTA
Add to Cart
Add to Cart

Includes

Vernier UVA Sensor, Built-in light diffuser, User manual

Requires

Requires an interface and software, sold separately

The UVA Sensor is an ultraviolet light sensor that responds primarily to UVA radiation (approximately 320 to 390 nm). This sensor is ideal for experiments using UV lamps.

UV peak sensitivity: one volt per 3940 mW/m2 at 340 nm Wavelength sensitivity region, approximate: 320 to 390 nm (320 to 375 nm, half-sensitivity points) Dimensions: 21 cm by 2 cm diameter Typical Resolution: 5 mW/m2 Time response: approximately 2 seconds to reach 95% of final reading Download the user manual for full specifications

Warranty and Services

5-year limited warranty

Specifications

Includes Built-in light diffuser
Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 5 mW/m2
Response Time Approximately 2 seconds to reach 95% of final reading
Dimensions (L x W) 21 cm length x 2 cm diameter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.