Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ UVB Sensor
SDPSCI_ED

Catalog No. S16273ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
Middle School, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16273ND Middle School, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16273ND Supplier Vernier™ Supplier No. UVBBTA
Add to Cart
Add to Cart

Includes

Vernier UVB Sensor, Built-in light diffuser, User manual

Requires

Requires an interface and software, sold separately

The UVB Sensor is an ultraviolet light sensor that responds primarily to UVB radiation (approximately 290 to 320 nm). It is ideal for experiments using sunlight as your UV source and is the sensor recommended for the UV experiments in our lab books.

UV peak sensitivity: one volt per 204 mW/m2 at 315 nm Wavelength sensitivity region, approximate: 290 to 320 nm, half-sensitivity points. Dimensions: 21 cm by 2 cm diameter Typical Resolution: 0.25 mW/m2 Time response: approximately 2 seconds to reach 95% of final reading Download the user manual for full specifications

Warranty and Services

5-year limited warranty

Specifications

Includes Built-in light diffuser
Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 0.25 mW/m2
Response Time Approximately 2 seconds to reach 95% of final reading
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.