Learn More
Eppendorf 3P UVETTE ADPTRW/20MM HT
Supplier:  Eppendorf 3P 4099005004
Encompass_Preferred
The Eppendorf UVette allows for measurements in the UV/Vis range with transparency from 220 nm to 1,600 nm with just a minimum volume of 50 µL. Choose between two optical path lengths (10 mm and 2 mm) with a single 90° rotation. The UVette is ideally matched for use with the Eppendorf BioPhotometer and Eppendorf BioSpectrometer. For use with other spectrophotometers, the UVette can be used with the aid of an adapter. • Aligning the UVette to the height of the light path • Panel function for optimal light path guidance
Catalog No. 50-118-0060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.