Learn More
Honeywell™ Uvex™ 880 Series Klip Lifts For Hard Hat Visors
Catalog No. 19048607 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Shade 3
Shade 6
Product Type:
880 Series Klip Lift, Full front
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Product Type
19-048607 Shade 6 880 Series Klip Lift, Full front
19-043653 Shade 3 880 Series Klip Lift, Full front
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-048607 Supplier Honeywell™ Supplier No. 328LFFB40000
  • $279.00
    $187.65 / Each of 1
    Save $91.35
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fits onto hard hat or bump cap brims, and worn in conjunction with existing protective eyewear. The Uvex 880 Series Klip Lifts for hard hat visors offer fast and convenient vision enhancement for a wide range of industries.
Whether you work in steelmaking, welding and cutting facilities, foundries or glassmaking operations, you will appreciate the convenience of the flip-up feature. These lenses can be quickly flipped down for observing a melt or a weld, and flipped back up again when not needed. Movement through a full 180° arc is possible, allowing the filter lenses to be raised or moved completely out of your line of vision.
  • Quickly flip down to enhance vision, then flip up when not needed
  • Integral spring clip holds 880 Series securely without screws, fasteners or tools
  • Available with half or full lenses in cobalt blue shades 3, 4 and 6
Specifications
Lens Tint Shade 6
Product Type 880 Series Klip Lift, Full front
For Use With (Equipment) Hardhat visors