Learn More
Honeywell™ Uvex™ Astrospec 3000™ Safety Glasses with Duoflex Temples
Catalog No. 189992554B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Blue
Red, White, Blue
Lens Tint:
Clear
Gray
SCT-Blue
Shade 2.0 Infra-dura
Silver Mirror
Lens Coating:
Anti-fog
Scratch-resistant
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
18-999-2554B Black Clear -
19-025334 Red, White, Blue Clear Anti-fog
17-985-30J Red, White, Blue Clear Scratch-resistant
18-999-2554A Black Clear Scratch-resistant
19-003-319 Black Shade 2.0 Infra-dura -
19-041-036 Black SCT-Blue Anti-fog
17-985-31J Red, White, Blue Gray Scratch-resistant
18-999-2553A Blue Clear -
18-999-2553B Blue Clear Anti-fog
18-999-2554D Black Silver Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-999-2554B Supplier Honeywell™ Supplier No. S1359C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Compliance
ANSI™ Z87+ and CSA Z94.3 standards
Specifications
Lens Type Uvextreme Anti-fog
Certifications/Compliance ANSI Z87+ and CSA Z94.3 standards
Lens Tint Clear
Frame Color Black
Temple Style Duoflex