Learn More
Honeywell™ Uvex Avatar™ Safety Glasses
Catalog No. 19039951
Change view
Click to view available options
Frame Color:
Black
Blue
Red
Teal
Lens Tint:
Amber
Clear
Gray
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-039-951 Blue Gray
19-039-953 Teal Clear
19-039-941 Black Clear
19-039-942 Black Gray
19-039-943 Black Amber
19-039-946 Red Clear
19-039-947 Red Gray
19-039-950 Blue Clear
19-039-954 Teal Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-039-951 Supplier Honeywell™ Supplier No. S2871HS
This item is not returnable. View return policy
Uvex Avatar™ is stylish and protective eyewear with technology allowing customizability, premium anti-fog coating and anti-scratch hard coating.
Polycarbonate Lens Coatings available in:
  • Hydroshield Anti-fog for 90X longer than the majority of other anti-fog products and 2X the scratch resistance than our next best anti-fog coating
  • Hardcoat
Fit:
  • Built with MMT—Multi-Material Technology providing a high level of adjustability
  • 9-base wrap-around lens provides uncompromised peripheral vision and protection
  • Ratchet lens: angle-adjustable temple length and lens inclination of 15 degrees
  • Soft, wire-core temples to customize to any facial profile
  • Flexible nose pads to fit any nasal profile and reduce slippage
  • Soft brow frame with indirect venting
  • Frames available in a variety of colors
Protection:
  • Certified to meet ANSI+ and tested by Honeywell to meet the requirements of the CSA Z94.3 standard
  • “U6” grade -level filtering >99.9% of UVB/UVA protection for far and near UV transmittance
  • Available in 4 frame colors and 5 lens tints
Style
  • ¾ frame, High performance, contemporary, stylish
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Type Hydroshield AF
Certifications/Compliance ANSI Z.87.1+,CSA Z94.3
Lens Tint Gray
Frame Color Blue
Temple Style Bayonette
Temple Color Black
UV Protection U6