Learn More
Description
Polycarbonate Lens Coatings available in:
- Hydroshield Anti-fog for 90X longer than the majority of other anti-fog products and 2X the scratch resistance than our next best anti-fog coating
- Hardcoat
Fit:
- Built with MMT—Multi-Material Technology providing a high level of adjustability
- 9-base wrap-around lens provides uncompromised peripheral vision and protection
- Ratchet lens: angle-adjustable temple length and lens inclination of 15 degrees
- Soft, wire-core temples to customize to any facial profile
- Flexible nose pads to fit any nasal profile and reduce slippage
- Soft brow frame with indirect venting
- Frames available in a variety of colors
Protection:
- Certified to meet ANSI+ and tested by Honeywell to meet the requirements of the CSA Z94.3 standard
- “U6” grade -level filtering >99.9% of UVB/UVA protection for far and near UV transmittance
- Available in 4 frame colors and 5 lens tints
Style
- ¾ frame, High performance, contemporary, stylish
Specifications
Specifications
| Lens Coating | Hardcoat |
| Certifications/Compliance | ANSI Z.87.1+,CSA Z94.3 |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| Frame Color | Red |
| Temple Style | Bayonette |
| Temple Color | Black |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.