Learn More
Honeywell™ Uvex™ Bionic™ Face Shield Accessories: Ratchet Suspension
Catalog No. 191305856 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Replacement Sweatband
Catalog No.
19-130-5856 Replacement Sweatband
1 options
Catalog No. 19-130-5856 Supplier Honeywell™ Supplier No. S8580
  • $19.01
    $11.65 / Each of 1
    Save $7.36
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Replacement sweatband for Uvex™ Bionic™ face shield is breathable, removable, and washable.
Uvex™ Bionic™ is a revolutionary face shield designed for rugged jobs. Extremely lightweight and highly adjustable for all-day comfort, it is perfect for workers exposed to impact, chemical splash, and airborne debris.
Specifications
Product Type Replacement Sweatband
For Use With (Equipment) Bionic Face Shields