Learn More
Honeywell™ Disposable Face Shields
Catalog No. 191300828 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Shade 5
Product Type:
Face Shield
Faceshield
Replacement visor
Lens Coating:
Anti-fog
Uncoated
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Product Type Lens Coating
19-130-0828 Clear Replacement visor -
19-130-0826 Clear Face Shield -
19-130-0827 Clear Faceshield Anti-fog
19-130-0831 Shade 5 Replacement visor Uncoated
19-130-0829 Clear Replacement visor Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-0828 Supplier Honeywell™ Supplier No. S8550
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for rugged jobs—tough yet lightweight and comfortable
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
  • Shell offers built-in chin protection and extended top-of-head coverage
  • Adjustable headgear tilts visor into 2784 possible positions
  • Ratcheting headgear features a locking mechanism for secure fit
  • Comfortable cell foam on back of headgear with breathable, removable, washable headband
  • 100% dielectric; no metal parts
Replacement Visors
  • Clear lens suits most indoor work applications
  • Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
Certifications/Compliance ANSI Z87.1-2010, CSA Z94.3
Lens Tint Clear
Product Type Replacement visor
For Use With (Equipment) For Bionic facesheild
Lens Material Polycarbonate