Learn More
Honeywell™ Uvex™ Protege™ Safety Eyewear with Hydroshield™ Anti-Fog Lens Coating
Catalog No. 19018529 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
SCT-Low IR
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-018-529 Gray
19-018-528 Clear
19-185-713 Amber
19-185-759 SCT-Low IR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-018-529 Supplier Honeywell™ Supplier No. S4201HS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Delivers anti-fog protection that lasts up to 60X longer
  • HydroShield™ anti-fog lens coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
Compliance
Meet ANSI™ Z87.1-2010 standards. Meet military V0 standard for impact protection
Specifications
Lens Type HydroShield AF
Lens Tint Gray
Frame Color Black