Learn More
Honeywell™ Uvex™ Carbonvision™ Safety Goggles
Catalog No. 19111204 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Material:
Fabric
Neoprene
Lens Tint:
Clear
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Material Lens Tint
19-111-204 Fabric Clear
19-111-203 Neoprene Gray
19-111-202 Neoprene Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-111-204 Supplier Honeywell™ Supplier No. S1650DF
  • $55.33
    $33.65 / Each of 1
    Save $21.68
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
Weighing in at just over an ounce and a half, Uvex Carbonvision goggles are one of the lightest wide vision goggles on the market. Now wearers can work for longer periods without the distractions and problems caused by discomfort, thereby increasing productivity and promoting a safer workplace.
Key Features and Benefits
  • Flexible soft frame adapts perfectly to the face to ensure a perfect fit and seal around the eyes for high-performance protection and all-day comfort
  • Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
  • Meets the ANSI Z87.1-2010 standard
  • Certified to the requirements of the CSA Z94.3 standard
For more information, visit www.fishersci.com and search for keywords UVEX Goggles.
Specifications
Lens Type Dura-streme HC/AF
Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety goggles
Frame Color Black, Gray
Frame Material Fabric