Learn More
Honeywell™ Uvex™ Classic™ Chemical Splash Closed-Vent Goggles
Catalog No. 19025406 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Closed vent model
Catalog No.
19-025406 Closed vent model
1 options
This item is not returnable. View return policy
Catalog No. 19-025406 Supplier Honeywell™ Supplier No. S364
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Lens Coating Anti-fog
Lens Type Uvextreme AF
Lens Tint Clear
Product Type Closed-vent Chemical Splash Goggles
For Use With (Equipment) Fits over most Rx spectacles
For Use With (Application) For dust, mist or chemical splash
Frame Color Clear