Learn More
Honeywell Uvex Clear™ Plus Towelettes
Catalog No. 19085493 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
19-085-493 Single-use
1 options
Catalog No. 19-085-493 Supplier Honeywell Supplier No. S470
  • $30.95
    $24.65 / Pack of 100
    Save $6.30
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $269.00
    $246.65 / Case of 10 PK
    Save $22.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Honeywell Uvex Clear™ Plus Towelettes swiftly remove dirt and debris
  • Anti-static lens towellete swiftly lifts and removes tough dirt, debris, and oily fingerprints
  • Soil reduced to below detectable limits and leave no haze, streaks, or residue behind
  • Improved formula delivers safe, effective results in an isopropanol- and silicone-free solution
  • Towelletes will not damage or delaminate lenses
  • Naturally scented with papaya extract
  • Pre-moistened, pocket-sized, and convenient for use on the go
  • 5 year shelf life
  • Simply wipe soiled lenses with the solution for ∼10 seconds and let dry to achieve a long-lasting clean
Specifications
Description Can be used at or above 40°C
Product Type Cleaning wipe
For Use With (Equipment) All Protective Eyewear
Dimensions (L x W x H) 2.25 in. x 5.25 in. x 8 in.
No. per Pack 100/Pk.
Silicone Free Yes
Material Paper
Disposable Single-use