Learn More
Honeywell™ Uvex™ Falcon Replacement Lenses
Description
Lighter than air, the Uvex Falcon soars above the competition as a great value in protective eyewear. The Uvex Falcon's frameless and optically perfected dual nine-base wrap-around lens provides superior coverage and unobstructed peripheral vision.
- Sleek, frameless design with wrap-around 9-base unilens
- Easy and economical lens replacement system
- Box of 10 clear replacement lenses with Ultra-dura Anti-scratch Coating
- Meets ANSI Z87.1-2003 (High Impact) and tested to meet the requirements of the CSA Z94.3 standard
Specifications
Specifications
| Lens Type | Ultradura |
| Description | Replacement lens |
| Lens Tint | Mirror |
| Product Type | Replacement lens |