Learn More
Honeywell™ Uvex™ Flex Seal™ Goggles
Exceptionally tight seal for protection from impacts and chemical splashes.
Supplier:  Honeywell™ S3400X
 View more versions of this product
Catalog No. 19-130-0832
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Description
Description
  • Contemporary
  • Low-profile
  • With wrap-around
  • Optically correct polycarbonate lens configuration increases visibility and protection
  • Uvextreme™ antifog coating and indirect ventilation minimize fogging
  • Temperature-resistant blue silicone frames fit over virtually all prescription eyewear
  • Pivoting clips permit goggle to be positioned on hardhat or on headband above the ears
Order Info
Available with neoprene band or fabric band.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Uvextreme Anti-fog
Safety goggles
Neoprene
Indirect
Silicone
Clear
Blue
Neoprene headband
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.