Learn More
Honeywell™ Uvex Flex Seal™ Safety Goggles
Catalog No. 19085466 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Headband Material:
Fabric
Neoprene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Headband Material
19-085-466 Neoprene
19-085-467 Fabric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-085-466 Supplier Honeywell™ Supplier No. S3400HS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Honeywell ™ Uvex Flex Seal™ Safety Goggles with ANSI, and CSA safety compliance
  • Lightweight, silicone body conforms to the face assuring the highest level of comfort and eye protection
  • Hydroshield™ AF coating provides 2x greater scratch resistance than other Honeywell™ anti-fog coatings
  • Uvextreme AF lens coating offers consistent fog-free time 90x longer than the majority of other anti-fog products
  • Tested by Honeywell™ to meet the requirements of the CSA Z94.3 standard
  • Silicone body stays supple even in extreme temperatures
  • Exceptional chemical splash and impact resistance
  • Fits over most prescription eyewear
  • Indirect venting system
  • Made in the USA
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87+ (High Impact), ANSI Z.87.1/1989, CA 21,199, CSA Z94.3
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) Abrasions, Airborne Particulate, Biohazard, Chemical, Impact, Splash
Frame Color Navy
Headband Material Neoprene
Includes Headband, Anti-Fog lens
No. per Case 50/Pk.
No. per Pack 1 Ea.
Size Universal
Ventilation Details Indirect
Weight (Metric) 0.3 kg
Lens Material Polycarbonate
Frame Material Silicone
UV Protection Uvextreme AF Anti-Fog Coating