Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Catalog No. 18813403 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Type:
Ultra-dura Scratch-resistant
Lens Tint:
Clear
Gray
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Type Lens Tint Magnification Power
18-813-403 Ultra-dura Scratch-resistant Gray +2.50
18-813-390 Ultra-dura Scratch-resistant Clear +1.00
18-813-402 Ultra-dura Scratch-resistant Gray +2.00
18-813-391 Ultra-dura Scratch-resistant Clear +1.50
18-813-392 Ultra-dura Scratch-resistant Clear +2.00
18-813-393 Ultra-dura Scratch-resistant Clear +2.50
18-813-394 Ultra-dura Scratch-resistant Clear +3.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 18-813-403 Supplier Honeywell™ Supplier No. S3773
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
High-impact protection
  • Soft, flexible nasal fingers
  • Padded temple tips and cushioned browbar
  • Adjustable temple length and ratcheting lens inclination provides for secure fit
  • Easy and economical lens replacement (see Cat. No. 18-813-424)
  • With Ultra-dura™ antiscratch coating
  • Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
Specifications
Lens Coating Scratch-resistant
Lens Type Ultra-dura Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2007
Lens Tint Gray
Product Type Reading Magnifier Spectacles
For Use With (Application) Minimizes outdoor sunlight and glare
Frame Color Black
Magnification Power +2.50
UV Protection 0.999