Learn More
Honeywell Uvex Genesis™ Safety Eyewear
Catalog No. 19185667 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Dark Gray
Catalog No. Lens Tint
19-185-667 Dark Gray
1 options
This item is not returnable. View return policy
Catalog No. 19-185-667 Supplier Honeywell Supplier No. S3212HS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to deliver all day comfort, protection and style. Built with Multi-Material technology and a high level of adjustability, Genesis provides workers with extraordinary comfort and raises the bar on compliance.
  • Wrap-around, 9-base uni-lens style for uncompromised peripheral vision and superior coverage
  • Soft elastomer browguard diffuses and deflects impact while providing comfort at all contact points
  • Soft flexible fingers on nose piece adjust and conform to almost any nasal profile to reduce slippage
  • Superior Hydroshield anti-fog coating keeps glasses consistently fog-free for 90X longer than the majority of other anti-fog products and 2X greater scratch resistance than our next best anti-fog coating
  • Adjustable temple length
  • Easy, economical lens replacement system
Specifications
Lens Coating Anti-fog
Lens Type Hydroshield Anti-Fog
Shape Wraparound
Lens Tint Dark Gray
Product Type Safety Glasses
Frame Color Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Half Frame