Learn More
Honeywell Safety Products™ Uvex™ Genesis™ Safety Eyewears
Catalog No. 19018521 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Espresso
SCT-Low IR
SCT-Vermilion
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-018-521 Espresso
19-018-520 Clear
19-185-742 SCT-Low IR
19-185-743 SCT-Vermilion
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-018-521 Supplier Honeywell Safety Products™ Supplier No. S3201HS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Wrap-around lens provides exceptional peripheral vision and protection
  • With Hydroshield™ anti-fog lens coating
  • Flexible fingers adjust to most nasal profiles
  • Elastomer browguard deflects impact
  • Adjustable temples and lens inclination
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
Specifications
Lens Type HydroShield AF
Lens Tint Espresso
Frame Color Black