Learn More
Honeywell™ Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Catalog No. 19042075
Change view
Click to view available options
Description:
Replacement Lenses/Spectacles
Replacement lens
Safety Spectacles
Lens Tint:
Amber
Clear
Gold Mirror
Gray (50%)
SCT-Low IR
Shade 3
Lens Coating:
Anti-fog
Hardcoat
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Lens Tint Lens Coating
19-042-075 Safety Spectacles Clear -
19-134-923 Replacement Lenses/Spectacles SCT-Low IR -
19-042-079 Safety Spectacles Gold Mirror Hardcoat
19-073-203 Replacement lens Gray (50%) Anti-fog
19-042-074 Safety Spectacles Clear -
19-042-078 Safety Spectacles Amber Hardcoat
19-148-321 Replacement Lenses/Spectacles Shade 3 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-042-075 Supplier Honeywell™ Supplier No. S6900X
This item is not returnable. View return policy
Wrap-around lens provides exceptional peripheral vision and protection. Flexible fingers adjust to most nasal profiles. Elastomer browguard deflects impact. Adjustable temples and lens inclination.
Uvex Genesis is engineered to deliver on the best all-day comfort, protection and style. It's built with MMT for comfort, and protects in a very wide assortment of lens tints. Uvex Genesis has an easy and economical lens replacement system. Frames have a lifetime guarantee. Made in the USA.
  • PC lens, Hydroshield AF, Dura-streme AF/HC, Uvextreme
  • Ratchet, adjustable temples, flexible nose, MMT
  • Available in a very wide range of lens tints
  • Economical lens replacement system
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Uvextreme Anti-fog
Description Safety Spectacles
Lens Tint Clear
Product Type Replacement lens
For Use With (Equipment) genesis Spectacles