Learn More
Honeywell™ Uvex™ Genesis™ Safety Glasses, Replacement Lenses
Description
Uvex Genesis is engineered to deliver on the best all-day comfort, protection and style. It's built with MMT for comfort, and protects in a very wide assortment of lens tints. Uvex Genesis has an easy and economical lens replacement system. Frames have a lifetime guarantee. Made in the USA.
- PC lens, Hydroshield AF, Dura-streme AF/HC, Uvextreme
- Ratchet, adjustable temples, flexible nose, MMT
- Available in a very wide range of lens tints
- Economical lens replacement system
Specifications
Specifications
| Lens Coating | Hardcoat |
| Lens Type | Ultradura hardcoat |
| Description | Safety Spectacles |
| Lens Tint | Gold Mirror |
| Product Type | Replacement lens |
| For Use With (Equipment) | genesis Spectacles |