Learn More
Honeywell Uvex Genesis™ XC Safety Eyewear
Catalog No. 19185673 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
SCT-Low IR
Shade 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-185-673 Shade 2
19-185-672 SCT-Low IR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-185-673 Supplier Honeywell Supplier No. S3305HS
  • $56.61
    $35.65 / Each
    Save $20.96
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $11,311.79
    $7,123.65 / Case of 200 EA
    Save $4,188.14
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Provides extended protection and more than 180 degrees of peripheral coverage. Uvex Genesis Eyewear world-renowned premium eyewear designed to deliver all day comfort, protection and style. Built with Multi-Material technology and a high level of adjustability, Genesis provides workers with extraordinary comfort and raises the bar on compliance.
  • High performance protection with extended wrap-around lens for extra cheek and side coverage
  • Soft elastomer browguard diffuses and deflects impact while providing comfort at all contact points
  • Soft flexible fingers on nose piece adjust and conform to almost any nasal profile to reduce slippage
  • Superior Hydroshield anti-fog coating keeps glasses consistently fog-free for 90X longer than the majority of other anti-fog products and 2X greater scratch resistance than our next best anti-fog coating
  • Adjustable ratcheting temples allow customized lens inclination and fit
  • Easy, economical lens replacement system
Specifications
Lens Coating Anti-fog
Lens Type Hydroshield Anti-Fog
Shape Wraparound
Lens Tint Shade 2
Product Type Safety Glasses
Frame Color Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Half Frame