Learn More
Honeywell™ Uvex™ Genesis™ XC Safety Eyewear
Catalog No. 19018525 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-018-525 Gray
19-018-524 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-018-525 Supplier Honeywell™ Supplier No. S3301HS
  • $425.00
    $266.65 / Pack of 10
    Save $158.35
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extended wrap-around lens for extra cheek and extra side protection
  • Soft flexible nose fingers conform to fit a variety of nasal profiles
  • Adjustable temples and lens inclination
  • HydroShield™ anti-fog lens coating
  • Provides 99.9% ultra-violet protection
  • Provides 2X greater scratch-resistance
Specifications
Lens Type HydroShield AF
Lens Tint Gray
Frame Color Black