Description
- Uvextreme Plus™ anti-fog lens coating with 10X longer-lasting anti-fog performance
- Also available with Dura-streme dual anti-fog/anti-scratch coating
- Foam-lined subframe
- Gap-free seal around eyes to protect from debris
- Flexible design can be worn with headband like goggle or with temples like spectacle
- Padded temples provide ideal tension and flex for comfort fit
- Ratcheting temples adjust lens angle by up to 15° for optimal coverage and air circulation
- Adjustable, flame-resistant latex-free cloth headband
- Wraparound frame design accommodates respirators, low-profile temples fits with earmuffs
- Wide lens tint assortment available in sealed eyewear: Clear, Amber, Espresso, Gray, SCT-Reflect 50 (indoor/outdoor mirror), SCT-Gray, SCT-Low IR, Shade 3.0 and Shade 5.0
- Available in Matte Black and Silver frame options
Certifications
CSA
Compliance
ANSI Z87.1-2010
Specifications
Specifications
| Lens Coating | Anti-static, Scratch-resistant, UV-treated |
| Lens Tint | Clear |
| Product Type | Sealed eyewear |
| For Use With (Application) | Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy |
| Frame Color | Matte Black |