Learn More
Honeywell™ Uvex™ Livewire™ Sealed Eyewear
Catalog No. 19039922
Change view
Click to view available options
Lens Tint:
Amber
Clear
Espresso
Gray
SCT-Gray Lens
SCT-Low IR
SCT-Reflect 50
Frame Color:
Black
Matte Black
Silver
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant, UV-treated
Anti-fog, Scratch-resistant
Anti-static, Scratch-resistant, UV-treated
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color Lens Coating
19-039-922 Espresso Matte Black Anti-static, Scratch-resistant, UV-treated
19-039-924 SCT-Reflect 50 Matte Black Anti-static, Scratch-resistant, UV-treated
19-185-763 Clear Black Anti-fog, Anti-static, Scratch-resistant, UV-treated
19-039-920 Clear Matte Black Anti-static, Scratch-resistant, UV-treated
19-039-921 Clear Matte Black Anti-fog, Scratch-resistant
19-039-923 Amber Matte Black Anti-static, Scratch-resistant, UV-treated
19-039-925 SCT-Gray Lens Matte Black Anti-static, Scratch-resistant, UV-treated
19-039-928 SCT-Low IR Matte Black Anti-static, Scratch-resistant, UV-treated
19-039-929 Clear Silver Anti-static, Scratch-resistant, UV-treated
19-039-930 Gray Silver Anti-static, Scratch-resistant, UV-treated
19-039-931 SCT-Reflect 50 Silver Anti-static, Scratch-resistant, UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 missing translation for 'options'
This item is not returnable. View return policy
Not for chemical splash applications
Catalog No. 19-039-922 missing translation for 'mfr' Honeywell™ missing translation for 'supplierNo' S2601XP
This item is not returnable. View return policy
Not for chemical splash applications
For protecting workers against intense wind, sun, dirt, dust, debris, as well as high heat and humidity
  • Uvextreme Plus™ anti-fog lens coating with 10X longer-lasting anti-fog performance
  • Also available with Dura-streme dual anti-fog/anti-scratch coating
  • Foam-lined subframe
  • Gap-free seal around eyes to protect from debris
  • Flexible design can be worn with headband like goggle or with temples like spectacle
  • Padded temples provide ideal tension and flex for comfort fit
  • Ratcheting temples adjust lens angle by up to 15° for optimal coverage and air circulation
  • Adjustable, flame-resistant latex-free cloth headband
  • Wraparound frame design accommodates respirators, low-profile temples fits with earmuffs
  • Wide lens tint assortment available in sealed eyewear: Clear, Amber, Espresso, Gray, SCT-Reflect 50 (indoor/outdoor mirror), SCT-Gray, SCT-Low IR, Shade 3.0 and Shade 5.0
  • Available in Matte Black and Silver frame options
Certifications
CSA
Compliance
ANSI Z87.1-2010
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-static, Scratch-resistant, UV-treated
Lens Tint Espresso
Product Type Sealed eyewear
For Use With (Application) Agriculture, Construction, Forestry, Manufacturing, Mining, Municipal Services, Oil and Gas, Steel and Metals, Utilities, Welding, Wind Energy
Frame Color Matte Black