Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Uvex™ Mercury™ Safety Glasses
Reagent Grade,≥99%
Supplier: Honeywell™ S1502
This item is not returnable.
View return policy
Description
- ≥95% for general laboratory use.

Specifications
Amber | |
Black, Gray |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction