Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Uvex™ Skyper™ Safety Glasses Replacement Lenses

Reagent Grade,≥99.0% (titration)

Specifications

Product Type Replacement lens
View More Specs

Products 5
Catalog Number Mfr. No. Lens Tint dimLensCoating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint dimLensCoating Price Quantity & Availability  
19-037-677
View Documents
Honeywell™
S6501X
Espresso N/A N/A N/A
19-038-475
View Documents
Honeywell™
S6505
SCT-reflect 50 N/A N/A N/A
19-038-313
View Documents
Honeywell™
S6507
Shade 3 N/A N/A N/A
19-038-314
View Documents
Honeywell™
S6508
Shade 5 N/A N/A N/A
19-037-674
View Documents
Honeywell™
S6500
Clear N/A N/A N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Specifications

Replacement lens
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.