Learn More
Honeywell Safety Products™ Uvex™ Skyper™ Safety Glasses
Catalog No. 19037656
Change view
Click to view available options
Lens Tint:
Clear
IR Filter Shade 2.0
IR Filter Shade 3.0
IR Filter Shade 5.0
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-037-656 Clear
19-038-311 IR Filter Shade 5.0
19-038-310 IR Filter Shade 3.0
19-038-309 IR Filter Shade 2.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-037-656 Supplier Honeywell Safety Products™ Supplier No. S1900
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Custom fit with superior peripheral protection
  • Polycarbonate lenses with single-lens design feature Ultradura™ hardcoat (UD) or Uvextreme™ AF (XTR) antifog, antiscratch, and antistatic coating
  • Wraparound, impact resistant lenses block 99.9% of harmful UVA and UVB radiation
  • Black frames with molded-in browguard reduce the danger of flying particulates
  • Molded-in nosebridge offers comfort and safety in a variety of work applications
  • Patented, adjustable, comfort-cushioned Duoflex™ temples
  • Simple lens replacement system, ratcheting lens inclination
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-1999 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Anti-static, Impact-resistant, Scratch-resistant
Shape Oval
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3-1999 standards.
Lens Tint Clear
Product Type Safety Glass
Frame Color Black
Material Polycarbonate (Lens)
Lens Material Polycarbonate