Learn More
Honeywell™ Uvex™ Stealth™ Chemical Splash Goggles Rx Carrier
Catalog No. 189994408 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Carrier
Catalog No.
18-999-4408 Carrier
1 options
Catalog No. 18-999-4408 Supplier Honeywell™ Supplier No. S3959
  • $44.75
    $22.65 / Each
    Save $22.10
    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $389.00
    $226.65 / Pack of 10 EA
    Save $162.35
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Adapter allows prescription glasses to be used with Uvex™ Stealth™ goggles. Carrier fits into center ventilation chamber of goggles.
Uvex™ Stealth™ goggles have a futuristic, low profile design for chemical splash and impact. Toric lens provides superior optics and peripheral vision. Quick adjust headband to create the highest levels of worker comfort and acceptance.
Specifications
Product Type Carrier
For Use With (Equipment) stealth Chemical Splash goggles
For Use With (Application) Allows prescription glasses to be used with Uvex™ Stealth™ goggles