Learn More
Honeywell™ Uvex™ Stealth™ OTG Goggles
Catalog No. 19185678 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Shade 5
Frame Material:
Elastomer
Fabric
Neoprene
Polycarbonate
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
For Use With (Application):
Cutting and gas welding operations
Ideal for Most Indoor Work Applications
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Material Lens Coating For Use With (Application)
19-185-678 Clear Elastomer Scratch-resistant Ideal for Most Indoor Work Applications
19-085-540 Clear Polycarbonate Anti-fog, Scratch-resistant -
18-813-410 Clear Fabric Anti-fog, Scratch-resistant Ideal for Most Indoor Work Applications
18-813-417 Shade 5 Neoprene Anti-fog, Scratch-resistant Cutting and gas welding operations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-185-678 Supplier Honeywell™ Supplier No. S3970HSF
Add to Cart
Add to Cart
Low-profile, wraparound style provides complete coverage designed for all-day wear
  • Ideal for impact, dust/airborne particles, chemical splash and optical radiation (UV and welding/heat radiation)
  • Superior optics and panoramic field of vision enhanced with toric lens
  • Pivoting clip and quick-adjusting wide headband
  • Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
  • Replacement lenses easily snap on
Order Info
Replacement parts and accessories available (see Cat. No.18-813-429).
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Lens Type Dura-streme Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type OTG Goggles
For Use With (Application) Ideal for Most Indoor Work Applications
Frame Color Navy
Headband Material Fabric
No. per Case 50/Cs.
Frame Material Elastomer