Learn More
Honeywell Uvex Stealth™ OTG Replacement Lens
Catalog No. 19185695 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Clear Lens
Catalog No.
19-185-695 Clear Lens
1 options
This item is not returnable. View return policy
Catalog No. 19-185-695 Supplier Honeywell Supplier No. S740HS
  • $38.62
    $24.65 / Each
    Save $13.97
    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $5,795.35
    $3,698.65 / Case of 150 EA
    Save $2,096.70
    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Economical and easy lens replacement allows for multiple lens tints, extending life of eyewear, increasing value and lowering cost of ownership. Lens meets impact resistance standards and provide superior optics and unobstructed peripheral vision.
  • Lens meets impact resistance standards and provide superior optics and unobstructed peripheral vision.
  • Superior Hydroshield anti-fog coating keeps glasses consistently fog-free for 90X longer than the majority of other anti-fog products and 2X greater scratch resistance than our next best anti-fog coating
Specifications
Lens Coating Anti-fog
Lens Type Hydroshield Anti-Fog
Lens Tint Clear
Product Type Replacement Lens
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
UV Protection UV400