Learn More
Honeywell™ Uvex™ Stealth™ Safety Reader Goggles
Catalog No. 19066153 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-066-153 +2.50
19-066-151 +1.50
19-066-152 +2.00
19-066-154 +3.00
19-066-150 +1.00
19-066-155 +1.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-066-153 Supplier Honeywell™ Supplier No. S3993X
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Uvex Stealth™ Reader goggle offers an extensive range of magnification strengths. With diopters from +1.0 to +3.0, the Uvex Stealth™ Reader goggle is ideal for workers who require both increased magnification for close-up inspection or fine detail work and the added protection of a goggle. Replacement lenses only.
The Uvex Stealth™ continues to be one of the most popular safety goggles on the market and represents the ultimate in modern design, technology and materials. We have again expanded the Uvex Stealth™ family of goggles to include reading magnifiers.
  • Available in Clear with Uvextreme™ anti-fog coating and five diopter strengths from +1.0 to +3.0
  • High quality, soft elastomer body conforms to facial contours for a secure, customized, gap-free fit
  • Goggle body is compatible with Uvex Stealth™ OTG lenses
  • Low profile, wrap-around style provides complete coverage and unobstructed vision complete coverage and protection from chemical splash
  • Wide, adjustable slide headband with pivoting clips provides an easy-to-adjust fit for complete comfort, even with hard hats
  • Economical lens replacement system extends the life of the goggle
  • Meets ANSI Z87.1-2010 (High Impact)
  • Tested by Honeywell to meet the requirements of the CSA Z94.3 standard
  • Made in USA
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Type Uv Extreme Anti-fog/Scratch-resistant
Width (Metric) 173 mm
Thickness (Metric) 2.55 mm
Lens Tint Clear
Product Type Safety Reader Goggles
Frame Color Blue
Magnification Power +2.50
Lens Material Polycarbonate
UV Protection 0.999