Learn More
Honeywell™ Uvex™ Strategy™ Safety Goggles
Catalog No. 19167204 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Gray
Orange
Includes:
Fabric band
Neoprene band
Ventilation Details:
Closed vent
Direct vent
Indirect vent
Indirect vent with foam
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Includes Ventilation Details
19-167-204 Gray Fabric band Indirect vent with foam
19-167-203 Gray Neoprene band Indirect vent
19-167-201 Gray Fabric band Direct vent
19-167-202 Gray Neoprene band Closed vent
19-167-205 Orange Neoprene band Indirect vent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-167-204 Supplier Honeywell™ Supplier No. S3815
  • $42.19
    $28.65 / Each of 1
    Save $13.54
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Economical over-the-glass (OTG) goggle that delivers both premium comfort and maximum protection
Provide protection against impact, chemical splash, dust, sand and debris — even in extreme environments. Available in four models for a variety of work applications: direct vent, closed vent, indirect vent and indirect vent with foam lining for fine particle protection.
  • Ideal for most indoor work applications
  • Rugged design that withstands the harshest environments
  • Lens tint: clear
  • Lenses offer 99.9% ultra-violet protection
  • Headband available in fabric or neoprene for chemical applications
  • Uvextra™ anti-fog coating minimizes fogging
  • Lightweight, TPE body provides exceptional comfort and a snug fit for maximum protection
  • Fits comfortably over most prescription eyewear
  • Economically priced
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog
Lens Type Uvextra
Lens Tint Clear
Product Type Safety goggles
Frame Color Gray
Includes Fabric band
Ventilation Details Indirect vent with foam