Learn More
Honeywell™ Uvex™ Tomcat™ Safety Glasses
Click to view available options
Lens Coating:
Impact-resistant
Lens Tint:
Clear
Gray
SCT-50
Silver Mirror
Description
- Optically corrected 10-base lens design
- Tight “wrap” style for peripheral protection with maximum visual clarity and quality
- Flexible rubber temples and adjustable saddle nosepads for custom fit
- Durable “gunmetal” frame
- Impact-resistant polycarbonate lenses block 99.9% of harmful UVA and UVB radiation
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Certifications/Compliance | Meet ANSI Z87.1-2003 standards. |
Lens Coating | Impact-resistant |
Lens Tint | Clear |
Frame Color | Gun Metal |
Temple Style | Rubber |
Lens Material | Polycarbonate |