Learn More
Honeywell™ Uvex™ Tomcat™ Safety Glasses
Sleek, wraparound metal frame design provides a sporty and stylish look
Supplier:  Honeywell™ S2453
 View more versions of this product
Catalog No. 19-050-914
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Optically corrected 10-base lens design
  • Tight “wrap” style for peripheral protection with maximum visual clarity and quality
  • Flexible rubber temples and adjustable saddle nosepads for custom fit
  • Durable “gunmetal” frame
  • Impact-resistant polycarbonate lenses block 99.9% of harmful UVA and UVB radiation
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Meet ANSI Z87.1-2003 standards.
Silver Mirror
Polycarbonate
Impact-resistant
Rubber
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.