Learn More
Honeywell™ Uvex™ XC™ Safety Glasses Replacement Lenses
Catalog No. 19066049
Change view
Click to view available options
Lens Tint:
Clear
Gray
SCT-Gray
SCT-Low IR
SCT-Reflect 50
Shade 2
Shade 5
Lens Type:
Ultradura
Uvextreme
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Type
19-066-049 SCT-Low IR Uvextreme
19-066-048 SCT-Gray Uvextreme
19-066-050 Shade 2 Ultradura
19-066-052 Shade 5 Ultradura
19-066-046 Gray Uvextreme
19-066-043 Clear Ultradura
19-066-047 SCT-Reflect 50 Ultradura
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-066-049 Supplier Honeywell™ Supplier No. S6954X
Extended wrap-around lens for extra cheek and extra side protection. Soft flexible nose fingers conform to fit a variety of nasal profiles. Adjustable temples and lens inclination.
Uvex Genesis XC has the same comfort and adjustability features as original Genesis, with 20% more extended wrap-around lens for extra cheek and side coverage. Rx lens insert available. Uvex Genesis XC has an easy and economical lens replacement system.
  • PC lens, Hydroshield AF, Dura-streme AF/HC
  • Available in a very wide range of lens tints
  • 20% more coverage than original Genesis
  • Economical lens replacement system
  • Meets military Vo ballistic impact requirement (on select tints)
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Uvextreme
Lens Tint SCT-Low IR
Product Type Replacement lens
For Use With (Equipment) XC Spectacles