Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Background Uniformity Screens

Catalog No. p-2622019 Shop All UVP Inc. Products
Click to view available options
Size:
13.1 x 9 .6 in.
18.68 x 12.95 in.

Specifications

For Use With (Application) Gel electrophoresis
Color Clear
Includes Background Uniformity Screen
For Use With (Equipment) UVP Transilluminators
Quantity each
Size 13.1 x 9 .6 in.
Product Type Background Uniformity Screen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.