Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP ChromaDoc-It™ TLC Imaging System

Catalog No. UVP97010901 Shop All UVP Inc. Products
Change view
Click to view available options
Camera Type:
DigiCam 130 digital color camera
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Camera Type
UVP97010901 DigiCam 130 digital color camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP97010901 Supplier UVP Supplier No. 97010901

missing translation for 'validMainframeMsg'

System with 5.1 megapixel digital color camera, viewing cabinet with overhead UV for TLC color documentation and imaging

  • High-intensity overhead 254/365nm UV plus white light
  • Doc-It software provides large selection of image capture and effects settings
  • Camera and capture software connect to your computer

Specifications

Voltage 115 V
Filter Type One Position
Height (English) 15.38 in.
Resolution 17.9 megapixel
Software Capture software enables live preview of images, user defined default settings and templates, annotations, use of filters, and report generation
Transilluminator 254 nm and 365 nm overhead UV; Interior white light
Camera Type DigiCam 130 digital color camera
Size L x W x H: 16.5 x 19.75 x 15.38 in.
For Use With (Application) Thin Layer Chromatography
Product Type ChromaDoc-It
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.