Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP Mineralight Shortwave UV Quartz Display Lamp

Catalog No. UVP95002607 Shop All UVP Inc. Products
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
UVP95002607 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP95002607 Supplier UVP Supplier No. 95002607
Only null left
Add to Cart
Add to Cart

Lamps have grid design, which is a highly uniform 254nm UV source of high-intensity

  • Lamp features a high intensity 254nm UV grid
  • UVP's special UVG shortwave filter reduces solarization, prolonging the filter life
  • Lamp mounts securely in its transformer base or is removable for movement around your work area
  • Lamp manufactured with rugged, scratch-resistant plastic
  • A snap-on filter assembly can be easily attached to convert this lamp for fluorescent analysis

Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis

Specifications

Voltage 115 V
Wavelength 254 nm
Case Material Plastic
Description UV grid display lamp, shortwave
Dimensions (L x W x H) 18.5 x 4.75 x 5 in. (470 x 121 x 127 mm)
For Use With (Application) Sterilization, Lapidary, Electronic Inspection
Height (English) 5 in.
Height (Metric) 127 mm
Includes UVG Long-LifeT Filter
Length (English) 18.5 in.
Length (Metric) 470 mm
Lighting Intensity 2200 mW/cm2 at 6 in.
Packaging Quantity 1 Ea.
Width (English) 4.75 in.
Width (Metric) 121 mm
Product Type UV Grid Display Lamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.