Learn More
Description
- This all-in-one system for gel documentation features an integrated 11.6 in. touch screen computer, a high resolution 6.3 MP camera - available as color or monochrome with an automated lens, a three position filter tray with pre-installed filters, overhead blue and white LED lighting and the UVP Elite UV Transilluminator.
- Easily switch between emission filters
- The three-position emission filter tray can be used to place three different filters and switch between them for different applications.
- A broad band filter (535 nm to 660 nm) is included with the system and a variety of other emission filters are available for additional purchase.
- Camera
- Choose from a 6.3MP color or monochrome camera with an automated lens.
- The Elite color camera can capture the natural color of the samples, eliminating the need for multiplexing and reducing post image processing for high quality color images.
- Intuitive Software with easy access to capture controls
- VisionWorks software on the UVP Solo Elite is a powerful yet intuitive software for capturing gel images and analyzing them.
- The simple user interface enables easy access to auto and manual image capture modes under the Device tab. Exposure time can be adjusted with customizable exposure settings.
- Captured images are saved in TIFF or JPEG formats.
- Extensive image enhancement and analysis tools
- Image enhancement and analysis features are included with all systems.
- Researchers can personalize their experiments and make use of enhancement features and annotation tools for publication purposes.
- The software offers many powerful tools such as noise reduction, histogram adjustment, inversion, pseudocolor, and more.
- The extensive analysis features are optimized for accurate and reproducible results.
- Several custom tools are available, such as automatic lane and band detection, 1D analysis, area density, and colony counting.
- Once the quantitation results have been generated, reports are created to show thorough analysis, such as Molecular Weight (MW), Rf, band intensities, and area density calculations.
- All the data can be conveniently exported to Excel.
Specifications
Specifications
| Voltage | 230 V |
| Filter Type | Ethidium Bromide |
| Resolution | 3072 x 2048 pixel |
| Software | VisionWorks |
| Transilluminator | UVP Elite UV Transilluminator |
| Lens Type | Automated |
| Camera Type | 6.3 MP Color |
| Dimensions | 61 x 46 x 38 cm |
| For Use With (Application) | Fluorescent DNA Gels, Fluorescent Protein Gels, Blue Light Gels, Colorimetry and More |
| Color | Black |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.