Learn More
UVP Universal Rocker ON/OFF Switch
Catalog No. 53012603 Shop All UVP Inc. Products
Change view
Click to view available options
Rocker; TLUM; B100
Catalog No.
53-012-603 Rocker; TLUM; B100
1 options
Catalog No. 53-012-603 Supplier UVP Supplier No. 53012603
Universal Rocker ON/OFF Switch for UVP Cabinets
Specifications
Product Type Rocker
For Use With (Equipment) B-100 high intensity UV lamps, Chromato-vue viewing cabinets, Transilluminators
Material Plastic
Voltage 115V
Includes Rocker Switch On/Off
No. per Pack 1 Ea.