Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UVP UVX Radiometer

Catalog No. UVP97001502 Shop All UVP Inc. Products
Change view
Click to view available options
Measures 254nm, 365nm or 302nm UV wavelengths
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
UVP97001502 Measures 254nm, 365nm or 302nm UV wavelengths
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UVP97001502 Supplier UVP Supplier No. 97001502

missing translation for 'validMainframeMsg'

Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths

  • The UVX's range switch provides selection from three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 microW/cm2 and 0 to 200 microW/cm2
  • A port for connecting a chart recorder (not included) allows for continuous monitoring
  • Conversion rate: 2.8 readings/second
  • Display: 3.5 digit LCD
  • Accuracy: ±2%
  • Sensors Accuracy: ±5% (includes NIST standard)
  • Dimensions: 6.2L x 3.6W x 2 in.H (157 x 91 x 51mm)

UVX Radiometer: Powered by a 9-volt battery for up to 120 hours
10:1 attenuator: Extends the reading range up to 200mW/cm2

Sensors sold separately.

Quality control

Specifications

Type UVP UVX Radiometer
For Use With (Application) Measure 254nm, 365nm or 302nm UV wavelengths
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.