Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Uvula Tissue Slides
SDP

Catalog No. NB151709 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151709 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151709 Supplier Novus Biologicals™ Supplier No. NBP311873

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Slides can be used with any antibody shown to be effective in formalin-fixed tissue using a standard immunostaining protocol.

In order to avoid false negative results from DNA or RNA sequences not being present in a 4 um cellular cross-section (NBP3-11873), 10 um tissue sections (NBP3-11874) are recommended. This product has not been evaluated for effectiveness in ISH or FISH applications.

Specifications

Quantity 5 Slides
Species Human
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Paraffin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.