Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ1.1 TCR Hamster anti-Mouse, Brilliant Violet 421, Clone: 2.11, BD

Catalog No. BDB566308 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566308 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566308 Supplier BD Biosciences Supplier No. 566308
Only null left
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The 2.11 monoclonal antibody recognizes the Vγ1.1 chain of the heterodimericγδT-cell receptor (TCRγδ) expressed by T lymphocytes and NK-T cells. It does not react with TCRαβ-bearing T cells. TCRγδassociates with CD3 molecules to form the TCRγδ/CD3 complex that mediates antigen recognition and can transduce intracellular signals leading to T cell responses. The Vγ1.1 TCR is also known as Vγ1 TCR. The Vγ1.1 variable region TCR gene segment primarily rearranges to join the Jγ4 and Cγ4 TCR gene segments. TCR Vγ1-expressing cells represent a major population of TCRγδcells in the adult mouse thymus, peripheral lymphoid tissues, and different epithelia. They constitute a minor population of TCRγδ-positive T cells during fetal and early postnatal life.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vγ1.1 TCR
Classification Monoclonal
Clone 2.11
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias TCR V gamma 1.1, TCR Vg1.1, TCR V-gamma-1, TCR Vg1, Cr4
Host Species Armenian Hamster
Immunogen Mouse TCR γδ+ T3.13.1 hybridoma cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.