Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100218 Supplier Invitrogen™ Supplier No. PA5100218
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATP6V1E1.

Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase E1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ATP6V1E1
Gene Accession No. P36543, P50518, Q6PCU2
Gene Alias 2410029D23Rik; Atp6e; Atp6e2; ATP6V1E; Atp6v1e1; ATPase H+ transporting V1 subunit E1; ATPase, H+ transporting lysosomal (vacuolar proton pump), 32 kDa; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1; ATPase, H+ transporting, lysosomal V1 subunit E1; ATPase, H+ transporting, V1 subunit E; D6Ertd385e; E2; H(+)-ATPase E-like protein; H(+)-transporting two-sector ATPase, 31kDa subunit; H+ ATPase subunit E; H+-transporting ATP synthase chain E, vacuolar; lysosomal 31kDa; P31; vacuolar H(+)-ATPase, E subunit; vacuolar H+ ATPase E1; vacuolar proton pump subunit E 1; V-ATPase 31 kDa subunit; V-ATPase subunit E; V-ATPase subunit E 1; VATPase, H+ transporting, lysosomal V1 subunit E1; V-ATPase, subunit E; Vma4; V-type proton ATPase subunit E 1
Gene Symbols ATP6V1E1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP6V1E1(Accession P36543), corresponding to amino acid residues K156-Q206.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11973, 297566, 529
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.