Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V-ATPase E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140273 Supplier Invitrogen™ Supplier No. PA5140273
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AKLRLSRIVE DPEVYQGLLD K Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 76%.

Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells. This isoform is essential for energy coupling involved in acidification of acrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase E2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP6V1E2
Gene Accession No. Q96A05
Gene Alias 4930500C14Rik; Atp6e1; ATP6EL2; Atp6v1e2; ATP6V1EL2; ATPase H+ transporting V1 subunit E2; ATPase, H transporting, lysosomal V1 subunit E2; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E-like 2; ATPase, H+ transporting, lysosomal V1 subunit E2; ATPase, H+ transporting, V1 subunit E-like 2; E1; lysosomal 31kDa; testis secretory sperm-binding protein Li 235P; vacuolar proton pump subunit E 2; vacuolar-type proton-translocating ATPase subunit E1; V-ATPase subunit E 2; VMA4; V-type proton ATPase subunit E 2
Gene Symbols ATP6V1E2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP6V1E2. Recombinant protein control fragment (Product #RP-109451).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.