Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V beta 5.1/5.2 Monoclonal Antibody (MR9-4), FITC, eBioscience™, Invitrogen™

Catalog No. p-7068224
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9117 25 μg
50-112-2137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-9117 Supplier Invitrogen™ Supplier No. 11579680
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide. Applications Reported: This MR9-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR9-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta 5.1/5.2
Applications Flow Cytometry
Classification Monoclonal
Clone MR9-4
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias TCR V beta5.1; TCR V beta5.2; TCRV beta 5.1; TCRV beta 5.2; TCRV beta5.1; TCRV beta5.2; Vb5.1; Vb5.2; Vbeta5.1; Vbeta5.2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.