Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-Gard™ Clear Polycarbonate Nitrometer PC Visor

Catalog No. 19141457 Shop All MSA Products
Change view
Click to view available options
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-141-457 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-141-457 Supplier MSA™ Supplier No. 10115837
Add to Cart
Add to Cart

Solution for general and basic applications and can be worn with helmet-mounted ear muffs. MSA™ V-Gard™ Clear Polycarbonate Nitrometer PC Visor reliably protects against impact, chemical splash and UltraViolet radiation.

  • Designed for use with V-Gard Frames and V-Gard Headgear
  • For many visor versions the sides are contoured towards face for a closer fit and improved resistance to impact and splash hazards
  • Can be worn with helmet-mounted ear muffs 
  • Superior UV protection
  • Special versions, called nitrometers, are designed to work withV-Gard Retractable or Standard Chin Protectors
  • Visors with green tint help alleviate eye strain and fatigue by reducing excessive glare, especially in outdoor working conditions. Not for use where a Shade Infrared (IR) visor is required.

Specifications

Color Clear
For Use With (Equipment) V-Gard Frame and Geadgear
Height (English) 8 in.
Height (Metric) 20.32 cm
Material Polycarbonate
Product Type General Purpose Visor
Quantity 1 Ea.
Style Contoured
Thickness 0.04 in. (1.02 mm)
Width (Metric) 43.18 cm
Width (English) 17 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.