Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-Gard™ H1 Safety Helmet Replacement Parts

Catalog No. 19001827 Shop All MSA Products
Change view
Click to view available options
Product Type:
Fas-Trac™ III Pivot Suspension
Nonvent/Bivent Chinstrap
Sweatband
Trivent Chinstrap
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-001-827 Nonvent/Bivent Chinstrap
19-001-824 Fas-Trac™ III Pivot Suspension
19-001-825 Sweatband
19-001-826 Trivent Chinstrap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-001-827 Supplier MSA™ Supplier No. 10194766
Add to Cart
Add to Cart

MSA™ V-Gard™ H1 Safety Helmet Replacement Parts help maintain the overall longevity of H1 Safety Helmets

  • Safety helmet designed for working at heights including tower climbing, forestry, rescue, and confined spaces
  • Replacement parts increase cost savings when new, full safety helmet is not required
  • Provides exceptional comfort and ease of use in a stylish low-profile hard hat design
  • A complete above-the-neck platform for all applications

Specifications

Certifications/Compliance ANSI/ISEA Z89.1-2014 TYPE 1, CLASS E (20,000 V ac) & CSA/CAN Z94.1-15 TYPE 1, CLASS E (20,000 V ac)
Color Black
Dimensions (L x W x H) 22 x 1.25 x 0.5 in.
For Use With (Equipment) V-Gard H1 Safety Helmets
For Use With (Application) Search and Rescue, Confined Space, Working at Heights, Turnarounds, Shutdowns, and Plant Outages
Material Polyester, Acetal
Product Type Nonvent/Bivent Chinstrap
Quantity 1 Replacement
Size Standard
Weight (English) 0.08 lb.
Head Style Front Brim (Short)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.