Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA V-Gard H2™ Replacement Sweatband, MIPS

Catalog No. 17200698 Shop All MSA Products
Change view
Click to view available options
Size:
6.5 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-698 6.5 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-698 Supplier MSA Supplier No. 10242631
Only null left
Add to Cart
Add to Cart

Replacement sweatband assembly for V-Gard H2™ helmets with MIPS.

  • Accessory

Specifications

Color Yellow
For Use With (Equipment) Hard Hats
Product Type Climbing Helmet
Size 6.5 to 8
Ventilated Vented
Head Style Front Brim
Width (Metric) 8.54 in.
Width (English) 8.54 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.